Please login to your online account to display your discounted pricing

Mouse anti-CD98, Clone: UM7F8, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD98
Clone UM7F8
Concentration 0.2mg/mL
Immunogen Molt 13 T cell line
Gene Accession No. P08195
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23429101

 
novus biologicals canada
NBP23429101ML
0.1mL Each for $139.00

NBP23429105

 
novus biologicals canada
NBP23429105ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD98
Clone UM7F8
Concentration 0.2mg/mL
Immunogen Molt 13 T cell line
Gene Accession No. P08195
Gene ID (Entrez) 6520
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting