Please login to your online account to display your discounted pricing

Mouse anti-CD98, Clone: UM7F8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23429105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23429105

  • / Each
 

Description & Specifications

Specifications

Antigen CD98
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P08195
Host Species Mouse
Immunogen Molt 13 T cell line
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 6520
Clone UM7F8
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Regulatory Status RUO
Primary or Secondary Primary