Please login to your online account to display your discounted pricing

Mouse anti-CD98, Clone: UM7F8, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450602

  • / Each
 

Description & Specifications

Specifications

Antigen CD98
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Molt 13 T cell line
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6520
Reconstitution Protein A purified
Concentration 1mg/mL
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08195
Isotype IgG1
Purification Method Purified
Regulatory Status RUO
Clone UM7F8
Conjugate Unlabeled