Please login to your online account to display your discounted pricing

Mouse anti-CD98, Clone: IPO-T10, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD98
Clone IPO-T10
Immunogen Native leucocytes
Gene Accession No. P08195
Gene ID (Entrez) 6520
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23446701

 
novus biologicals canada
NBP23446701ML
0.1mL Each for $139.00

NBP23446705

 
novus biologicals canada
NBP23446705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD98
Clone IPO-T10
Immunogen Native leucocytes
Gene Accession No. P08195
Gene ID (Entrez) 6520
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgM
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)