Please login to your online account to display your discounted pricing

Mouse anti-CD98, Clone: IPO-T10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446701

  • / Each
 

Description & Specifications

Specifications

Antigen CD98
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Immunogen Native leucocytes
Purification Method Concentrated
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 6520
Cross Reactivity Human
Gene Accession No. P08195
Host Species Mouse
Isotype IgM
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone IPO-T10
Conjugate Unlabeled