Please login to your online account to display your discounted pricing

Mouse anti-CD98, Clone: IPO-T10, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23446705ML

 View all options for this product


For Research Use Only

Catalog No. NBP23446705


Description & Specifications


Antigen CD98
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Clone IPO-T10
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P08195
Host Species Mouse
Immunogen Native leucocytes
Isotype IgM
Purification Method Concentrated
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6520