missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD98, Clone: 1C11.7E3, Novus Biologicals™

$109.00 - $259.02

Specifications

Antigen CD98
Clone 1C11.7E3
Concentration 1.0mg/mL
Immunogen CD98
Gene ID (Entrez) 6520
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP236491 Novus Biologicals Canada
NBP236491
0.1mg Each for $259.02
NBP236491SS Novus Biologicals Canada
NBP236491SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

CD98
1.0mg/mL
6520
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Unlabeled
Purified
1C11.7E3
CD98
PBS. Protein G purified.
Primary
Murine
Purified
Flow Cytometry,Western Blot