missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD98, Clone: 1C11.7E3, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP236491SS

 View more versions of this product

Catalog No. NBP236491SS


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

CD98
1C11.7E3
Unlabeled
PBS. Protein G purified.
CD98
0.025mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Flow Cytometry,Western Blot
1.0mg/mL
Purified
Murine
Purified
RUO
Primary
6520