missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD90/Thy1, Clone: F15-42-1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD90/Thy1
Clone F15-42-1
Concentration 1mg/mL
Immunogen Purified human brain Thy1
Gene Accession No. P04216
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23331601 Novus Biologicals Canada
NBP23331601MG
0.1mg Each for $384.00
NBP23331602 Novus Biologicals Canada
NBP23331602MG
0.2mg Each for $527.00
NBP23461501 Novus Biologicals Canada
NBP23461501MG
0.1mg Each for $384.00
NBP23461502 Novus Biologicals Canada
NBP23461502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD90/Thy1
1mg/mL
P04216
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
F15-42-1
Purified human brain Thy1
7070
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Functional Assay,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation