Please login to your online account to display your discounted pricing

Mouse anti-CD90/Thy1, Clone: F15-42-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461501

  • / Each
 

Description & Specifications

Specifications

Antigen CD90/Thy1
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7070
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04216
Host Species Mouse
Immunogen Purified human brain Thy1
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone F15-42-1
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Primate