Please login to your online account to display your discounted pricing

Mouse anti-CD90/Thy1, Clone: F15-42-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461502

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Gene Accession No. P04216
Purification Method Purified
Cross Reactivity Human
Cross Reactivity Primate
Regulatory Status RUO
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene ID (Entrez) 7070
Quantity 0.2mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Primary or Secondary Primary
Immunogen Purified human brain Thy1
Antigen CD90/Thy1
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Clone F15-42-1
Reconstitution Protein A purified
Isotype IgG1, κ