Please login to your online account to display your discounted pricing

Mouse anti-CD90/Thy1, Clone: F15-42-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23331602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23331602

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Antigen CD90/Thy1
Format Purified
Gene Accession No. P04216
Quantity 0.2mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Primate
Formulation 1.0mg/mL of Ab purified from Bioreactor Concentrate by Protein A/G Prepared in 10mM PBS. IgG purified.
Host Species Mouse
Immunogen Purified human brain Thy1.
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 7070
Clone F15-42-1
Concentration 0.2mg/mL
Conjugate Unlabeled