Please login to your online account to display your discounted pricing

Mouse anti-CD90/Thy1, Clone: 5E10, Novus Biologicals™

$274.00 - $329.00

Specifications

Antigen CD90/Thy1
Clone 5E10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD90.
Gene Accession No. P04216
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14337901

 
novus biologicals canada
NBP14337901MG
0.1mg Each for $329.00

NBP14337903

 
novus biologicals canada
NBP1433790025MG
0.025mg Each for $274.00
Description & Specifications

Specifications

Antigen CD90/Thy1
Clone 5E10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD90.
Gene Accession No. P04216
Gene ID (Entrez) 7070
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting