Please login to your online account to display your discounted pricing

Mouse anti-CD90/Thy1, Clone: 5E10, Cy5.5, PerCP, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen CD90/Thy1
Clone 5E10
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD90.
Gene Accession No. P04216
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP142847

 
novus biologicals canada
NBP142847100TESTS
100 Tests Each for $395.00

NBP14284725

 
novus biologicals canada
NBP14284725TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen CD90/Thy1
Clone 5E10
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD90.
Gene Accession No. P04216
Gene ID (Entrez) 7070
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Cy5.5, PerCP
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry