Please login to your online account to display your discounted pricing

Mouse anti-CD9, Clone: SN4, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD9
Clone SN4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14344401

 
novus biologicals canada
NBP14344401MG
0.1mg Each for $285.00

NBP14344403

 
novus biologicals canada
NBP1434440025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen CD9
Clone SN4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation