Please login to your online account to display your discounted pricing

Mouse anti-CD9, Clone: SN4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434440025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344403

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Antigen CD9
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Clone SN4
Formulation PBS [pH 7.2]
Host Species Mouse
Immunogen The immunogen for this antibody was CD9.
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 928
Conjugate Unlabeled
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Cross Reactivity Human