Please login to your online account to display your discounted pricing

Mouse anti-CD9, Clone: SN4, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD9
Clone SN4
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200047

 
novus biologicals canada
NBP200047100TESTS
100 Tests Each for $252.00

NBP20004725

 
novus biologicals canada
NBP20004725TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD9
Clone SN4
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry