Please login to your online account to display your discounted pricing

Mouse anti-CD9, Clone: SN4/C3-3A2, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CD9
Clone SN4/C3-3A2
Concentration 0.2mg/mL
Immunogen Native purified CD9 from NALM-6 (human pre-B leukemia cell line).
Gene ID (Entrez) 928
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143893

 
novus biologicals canada
NBP143893100TESTS
100 Tests Each for $296.00

NBP14389325

 
novus biologicals canada
NBP14389325TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD9
Clone SN4/C3-3A2
Concentration 0.2mg/mL
Immunogen Native purified CD9 from NALM-6 (human pre-B leukemia cell line).
Gene ID (Entrez) 928
Formulation PBS [pH 7.2] containing BSA and/or gelatin for protein stabilization
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry