Please login to your online account to display your discounted pricing

Mouse anti-CD9, Clone: SN4, Biotin, Novus Biologicals™

$131.00 - $197.00

Specifications

Antigen CD9
Clone SN4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14368201

 
novus biologicals canada
NBP14368201MG
0.1mg Each for $197.00

NBP14368203

 
novus biologicals canada
NBP1436820025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen CD9
Clone SN4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry