Please login to your online account to display your discounted pricing

Mouse anti-CD83, Clone: HB15e, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD83
Clone HB15e
Immunogen The immunogen was the full length version of the protein: CD83 (HB15e)
Gene ID (Entrez) 9308
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14361401

 
novus biologicals canada
NBP14361401MG
0.1mg Each for $252.00

NBP14361403

 
novus biologicals canada
NBP1436140025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen CD83
Clone HB15e
Immunogen The immunogen was the full length version of the protein: CD83 (HB15e)
Gene ID (Entrez) 9308
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry