missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD8 beta, Clone: 341, FITC, Novus Biologicals™

$175.00 - $505.00

Specifications

Antigen CD8 beta
Clone 341
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD8b.
Gene ID (Entrez) 926
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP1440395M Novus Biologicals Canada
NBP144039005MG
0.05mg Each for $175.00
NBP1440395 Novus Biologicals Canada
NBP14403905MG
0.5mg Each for $505.00
Specifications Description & Specifications

Specifications

CD8 beta
0.5mg/mL
926
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Affinity Purified
Flow Cytometry
341
The immunogen for this antibody was CD8b.
PBS [pH 7.2]
Primary
Murine
FITC
Affinity Purified