Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: SPM548, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23283510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23283510

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Gene Accession No. P01732
Antigen CD8 alpha
Host Species Mouse
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Formulation PBS with 0.05% BSA. Protein G purified.
Immunogen Human CD8 recombinant protein
Purification Method Purified
Quantity 1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 925
Clone SPM548
Concentration 0.2mg/mL