missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: SPM548, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD8 alpha
Clone SPM548
Concentration 1mg/mL
Immunogen Human CD8 recombinant protein
Gene Accession No. P01732
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23479002 Novus Biologicals Canada
NBP23479002MG
0.2mg Each for $527.00
NBP23479001 Novus Biologicals Canada
NBP23479001MG
0.1mg Each for $384.00
Specifications Description & Specifications

Specifications

CD8 alpha
1mg/mL
P01732
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM548
Human CD8 recombinant protein
925
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot