Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: SPM548, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Immunogen Human CD8 recombinant protein
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Antigen CD8 alpha
Concentration 1mg/mL
Isotype IgG1
Conjugate Unlabeled
Quantity 0.1mg
Clone SPM548
Reconstitution Protein A purified
Primary or Secondary Primary
Gene ID (Entrez) 925
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Format Purified
Gene Accession No. P01732
Purification Method Purified