Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: HIT8a, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CD8 alpha
Clone HIT8a
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD8.
Gene ID (Entrez) 925
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143895

 
novus biologicals canada
NBP143895100TESTS
100 Tests Each for $296.00

NBP14389525

 
novus biologicals canada
NBP14389525TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD8 alpha
Clone HIT8a
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD8.
Gene ID (Entrez) 925
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry