Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: HIT8a, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD8 alpha
Clone HIT8a
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD8.
Gene ID (Entrez) 925
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14344601

 
novus biologicals canada
NBP14344601MG
0.1mg Each for $285.00

NBP14344603

 
novus biologicals canada
NBP1434460025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen CD8 alpha
Clone HIT8a
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD8.
Gene ID (Entrez) 925
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)