Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: HIT8a, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14344601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344601

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Monoclonal or Polyclonal Monoclonal
Antigen CD8 alpha
Primary or Secondary Primary
Gene ID (Entrez) 925
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was CD8.
Conjugate Unlabeled
Format Affinity Purified
Regulatory Status RUO
Quantity 0.1mg
Cross Reactivity Human
Host Species Mouse
Concentration 0.5mg/mL
Clone HIT8a
Isotype IgG1, κ
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified