Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: HIT8a, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434460025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344603

  • / Each
 

Description & Specifications

Specifications

Immunogen The immunogen for this antibody was CD8.
Quantity 0.025mg
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Cross Reactivity Human
Storage Requirements Store at 4°C. Do not freeze.
Formulation Phosphate buffer pH 7.2
Format Affinity Purified
Primary or Secondary Primary
Antigen CD8 alpha
Isotype IgG1, κ
Host Species Mouse
Regulatory Status RUO
Clone HIT8a
Gene ID (Entrez) 925
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Purification Method Affinity Purified