Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: C8/468, Novus Biologicals™

$139.00 - $139.00

Specifications

Antigen CD8 alpha
Clone C8/468
Concentration 0.2mg/mL
Immunogen Human CD8 recombinant protein
Gene Accession No. P01732
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP22942601

 
novus biologicals canada
NBP22942601ML
0.1mL Each for $139.00

NBP23295201

 
novus biologicals canada
NBP23295201ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen CD8 alpha
Clone C8/468
Concentration 0.2mg/mL
Immunogen Human CD8 recombinant protein
Gene Accession No. P01732
Gene ID (Entrez) 925
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting