Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: C8/468, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP22942601ML

 View all options for this product


For Research Use Only

Catalog No. NBP22942601


Description & Specifications


Antigen CD8 alpha
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone C8/468
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P01732
Host Species Mouse
Immunogen A recombinant C-terminus protein fragment specific to CD8 was used as the immunogen for the C8/468 antibody.
Isotype IgG1, κ
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 925
Test Specificity CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas are CD4+/CD8-. Anaplastic large cell lymphoma is usually CD4+ and CD8-, and in T-lymphoblastic lymphoma/leukemia, CD4 and CD8 are often co-expressed. CD8 is also found in littoral cell angioma of the spleen.