Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: C8/468 + C8/144B, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23431805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23431805

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen Human CD8 recombinant protein (C8/468); A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule (C8/144B)
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 925
Reconstitution Protein A purified
Clone C8/468 + C8/144B
Conjugate Unlabeled
Antigen CD8 alpha
Gene Accession No. P01732
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL
Cross Reactivity Human