Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: C8/468, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen CD8 alpha
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Human CD8 recombinant protein
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 925
Conjugate Unlabeled
Gene Accession No. P01732
Isotype IgG1
Purification Method Purified
Reconstitution Protein A purified
Clone C8/468
Concentration 1mg/mL