Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: C8/144B, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD8 alpha
Clone C8/144B
Concentration 1mg/mL
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule.
Gene Accession No. P01732
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23458801

 
novus biologicals canada
NBP23458801MG
0.1mg Each for $384.00

NBP23458802

 
novus biologicals canada
NBP23458802MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD8 alpha
Clone C8/144B
Concentration 1mg/mL
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule.
Gene Accession No. P01732
Gene ID (Entrez) 925
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting