Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: C8/144B, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458802

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen CD8 alpha
Format Purified
Gene Accession No. P01732
Host Species Mouse
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule.
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone C8/144B
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 925
Reconstitution Protein A purified
Concentration 1mg/mL