Please login to your online account to display your discounted pricing

Mouse anti-CD8 alpha, Clone: C8/144B, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458801

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Quantity 0.1mg
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule.
Conjugate Unlabeled
Cross Reactivity Human
Primary or Secondary Primary
Isotype IgG1
Format Purified
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Gene ID (Entrez) 925
Reconstitution Protein A purified
Clone C8/144B
Gene Accession No. P01732
Purification Method Purified
Antigen CD8 alpha