Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: SPM550, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD79A
Clone SPM550
Concentration 0.2mg/mL
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Gene Accession No. P11912
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23283401

 
novus biologicals canada
NBP23283401ML
0.1mL Each for $139.00

NBP23283410

 
novus biologicals canada
NBP23283410ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CD79A
Clone SPM550
Concentration 0.2mg/mL
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Gene Accession No. P11912
Gene ID (Entrez) 973
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting