Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: SPM550, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD79A
Clone SPM550
Concentration 1mg/mL
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Gene Accession No. P11912
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23479201

 
novus biologicals canada
NBP23479201MG
0.1mg Each for $384.00

NBP23479202

 
novus biologicals canada
NBP23479202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD79A
Clone SPM550
Concentration 1mg/mL
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Gene Accession No. P11912
Gene ID (Entrez) 973
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot