Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: SPM550, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479202

  • / Each
 

Description & Specifications

Specifications

Antigen CD79A
Format Purified
Gene ID (Entrez) 973
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P11912
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone SPM550
Concentration 1mg/mL
Conjugate Unlabeled