Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: SPM550, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479201

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Format Purified
Conjugate Unlabeled
Antigen CD79A
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1, κ
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Gene Accession No. P11912
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Host Species Mouse
Gene ID (Entrez) 973
Purification Method Purified
Concentration 1mg/mL
Clone SPM550
Reconstitution Protein A purified