Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: SPM549, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23283310ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23283310

  • / Each
 

Description & Specifications

Specifications

Antigen CD79A
Format Purified
Gene Accession No. P11912
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 973
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Conjugate Unlabeled
Clone SPM549
Concentration 0.2mg/mL