Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: SPM549, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23283301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23283301

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 973
Cross Reactivity Human
Antigen CD79A
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P11912
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 0.2mg/mL
Clone SPM549
Conjugate Unlabeled