Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: SPM549, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479102

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Cross Reactivity Human
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Format Purified
Conjugate Unlabeled
Antigen CD79A
Concentration 1mg/mL
Isotype IgG1, κ
Gene ID (Entrez) 973
Gene Accession No. P11912
Clone SPM549
Purification Method Purified
Reconstitution Protein A purified