Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: JCB117 + HM47/A9, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD79A
Clone JCB117 + HM47/A9
Concentration 0.2mg/mL
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein (JCB117 and HM47/A9).
Gene Accession No. P11912
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23433005

 
novus biologicals canada
NBP23433005ML
0.5mL Each for $321.00

NBP23433001

 
novus biologicals canada
NBP23433001ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen CD79A
Clone JCB117 + HM47/A9
Concentration 0.2mg/mL
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein (JCB117 and HM47/A9).
Gene Accession No. P11912
Gene ID (Entrez) 973
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting