Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: JCB117 + HM47/A9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433005

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD79A
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein (JCB117 and HM47/A9).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Gene Accession No. P11912
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 973
Reconstitution Protein A purified
Clone JCB117 + HM47/A9
Concentration 0.2mg/mL
Conjugate Unlabeled
Quantity 0.5mL