Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: JCB117 + HM47/A9, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD79A
Clone JCB117 + HM47/A9
Concentration 1mg/mL
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein (JCB117 and HM47/A9).
Gene Accession No. P11912
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23465301

 
novus biologicals canada
NBP23465301MG
0.1mg Each for $384.00

NBP23465302

 
novus biologicals canada
NBP23465302MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD79A
Clone JCB117 + HM47/A9
Concentration 1mg/mL
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein (JCB117 and HM47/A9).
Gene Accession No. P11912
Gene ID (Entrez) 973
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot