Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: JCB117 + HM47/A9, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465302

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Conjugate Unlabeled
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein (JCB117 and HM47/A9).
Isotype IgG1, κ
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD79A
Format Purified
Purification Method Purified
Concentration 1mg/mL
Regulatory Status RUO
Quantity 0.2mg
Gene ID (Entrez) 973
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P11912
Monoclonal or Polyclonal Monoclonal
Clone JCB117 + HM47/A9