Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: JCB117, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465202

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Gene ID (Entrez) 973
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Cross Reactivity Human
Primary or Secondary Primary
Quantity 0.2mg
Reconstitution Protein A purified
Gene Accession No. P11912
Purification Method Purified
Isotype IgG1, κ
Concentration 1mg/mL
Format Purified
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Clone JCB117
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Antigen CD79A