Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: JCB117, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD79A
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P11912
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 973
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Protein A purified
Clone JCB117