Please login to your online account to display your discounted pricing

Mouse anti-CD79A, Clone: HM47/A9, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD79A
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Host Species Mouse
Gene Accession No. P11912
Format Purified
Immunogen A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Concentration 1mg/mL
Regulatory Status RUO
Conjugate Unlabeled
Clone HM47/A9
Purification Method Purified
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 973
Isotype IgG1, κ
Reconstitution Protein A purified