Please login to your online account to display your discounted pricing

Mouse anti-CD74, Clone: SPM523, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD74
Clone SPM523
Concentration 0.2mg/mL
Immunogen SU-DHL-4 lymphoma cells
Gene Accession No. P044233
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23294701

 
novus biologicals canada
NBP23294701ML
0.1mL Each for $139.00

NBP23294710

 
novus biologicals canada
NBP23294710ML
1mL Each for $429.00
Description & Specifications

Specifications

Antigen CD74
Clone SPM523
Concentration 0.2mg/mL
Immunogen SU-DHL-4 lymphoma cells
Gene Accession No. P044233
Gene ID (Entrez) 972
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting